missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11978-2-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors.Spezifikation
GNB4 | |
Polyclonal | |
Unconjugated | |
GNB4 | |
GNB4, Transducin beta chain 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14696, 294962, 59345 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35353, P29387, Q9HAV0 | |
GNB4 | |
GNB4 Fusion Protein Ag2604 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |