Learn More
Gibco™ TGFB2 Recombinant Human Protein
Recombinant Human Transforming Growth Factor β 2 (TGFB2) is a bioactive protein intended for use in cell culture applications
Marke: Gibco™ PHG9114
Additional Details : Gewicht : 0.02000kg
Beschreibung
TGFB2 binds the transforming growth factor-β receptor and plays a role in regulation of cell growth and proliferation. It may be involved in mesenchymal-epithelial cell interactions during development.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by its ability to inhibit the mouse IL-4 dependent proliferation of mouse HT-2 cells
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Spezifikation
Cell Culture | |
25 kDa | |
5μg | |
Human | |
Store in freezer (-5 to -30°C). | |
5 μg | |
Baculovirus | |
Recombinant, Ligand | |
TGFs (Transforming Growth Factors) | |
Gibco™ |
7042 | |
Sequential Chromatography | |
TGF-β2 | |
Recombinant | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Oncology, Stem Cell Research, Cardiovascular Research, Differentiation, Bone Research | |
98 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.