missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11941-2-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Gas2 is a 313 amino acid protein encoded by the human gene GAS2. Gas2 is thought to play a role in apoptosis by acting as a cell death substrate for caspases. Gas2, a component of the microfilament system, is cleaved by a caspase (caspase-3 and caspase-7) at Asparagine 278 during apoptosis. The cleaved form resulting from this dramatically induces the rearrangement of the Actin cytoskeleton and causes potent changes in the shape of the affected cells. Gas2 is believed to also be involved in the membrane ruffling process. During the G0-G1 transition phase Gas2 can be found phosphorylated on its serine residues. Gas2 is a cytoskeleton and peripheral membrane protein that co-localizes with Actin fibers at the cell border and along the stress fibers in growth-arrested fibroblasts. Gas2 is mainly membrane-associated but when hyperphosphorylated it will accumulate at membrane ruffles. Gas2 is specifically expressed at growth arrest and is ubiquitously expressed with highest levels found in liver, lung and kidney.Spezifikation
GAS2 | |
Polyclonal | |
Unconjugated | |
GAS2 | |
GAS 2, GAS2, growth arrest 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14453, 2620, 499156 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43903, P11862 | |
Gas2 | |
GAS2 Fusion Protein Ag2540 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |