missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GARP Mouse, APC, Clone: GARP5, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio QAB84-APC-100Tests
Additional Details : Gewicht : 0.01000kg
Spezifikation
GARP | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
LRRC32 | |
Protein A/G Purified | |
RUO | |
2615 | |
IgG1 |
Flow Cytometry | |
GARP5 | |
APC | |
XP_016873016 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |