missing translation for 'onlineSavingsMsg'
Learn More

Gabpb2 Antibody, Novus Biologicals™

Artikelnummer. 18267026 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
25ul
Packungsgröße:
0.10 Milliliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18267026 0.1 mL 0.10 Milliliter
18419350 25ul 25 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18267026 Lieferant Novus Biologicals Lieferanten-Nr. NBP181132

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Gabpb2 Polyclonal specifically detects Gabpb2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifica

Antigen Gabpb2
Anwendungen Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Zusammensetzung PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gen-Zugriffsnummer Q8TAK5
Gen-Alias GA binding protein transcription factor, beta subunit 2, GA-binding protein subunit beta-2, GABP subunit beta-2, GABPB-2, MGC29891
Gensymbole GABPB2
Wirtsspezies Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:DKSAFDIALEKNNAEILVILQEAMQNQVNVNPERANPVTDPVSMAAPFIFTSGEVVNLASLISSTNTKTTSGDPHASTVQFSNSTTS
Reinigungsverfahren Affinity Purified
Menge 0.1 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 126626
Testspezifität Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.