missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FYN A, GST Tag Recombinant Protein

Human FYN A, GST Tag Recombinant Protein

Marke:  Invitrogen™ PV6347

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15438983

  • 1942.00 CHF / 100 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Fyn has been shown to phosphorylate Dab1, an intracellular adaptor protein that interacts with amyloid precursor protein (APP) and apoE receptor 2 (apoEr2). The interaction of Fyn and Dab1 regulates the phosphorylation, trafficking, and processing of APP and apoEr2. Fyn expression has been shown to be significantly increased in Chronic Myelogenous Leukemia (CML). Knockdown of Fyn with shRNA slows leukemia cell growth, inhibits clonogenicity, and leads to increased sensitivity to imatinib.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Spezifikation

Spezifikation

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
87.1kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
100μg
SLK, SYN, MGC45350
Recombinant
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote