missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FYN A, GST Tag Recombinant Protein
Human FYN A, GST Tag Recombinant Protein
574.00 CHF - 8710.00 CHF
Spezifikation
Gen-Alias | SLK, SYN, MGC45350 |
---|---|
Spezies | Human |
Rekombinant | Recombinant |
Proteinmarkierung | GST Tag |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15428983
|
Invitrogen™
PV6346 |
10μg |
574.00 CHF
10 Mikrogramm |
Verfügbar ab: 26-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15438983
|
Invitrogen™
PV6347 |
100μg |
1942.00 CHF
100 Mikrogramm |
Verfügbar ab: 26-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15448983
|
Invitrogen™
PV6348 |
1mg |
8710.00 CHF
1 Milligramm |
Verfügbar ab: 26-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Fyn has been shown to phosphorylate Dab1, an intracellular adaptor protein that interacts with amyloid precursor protein (APP) and apoE receptor 2 (apoEr2). The interaction of Fyn and Dab1 regulates the phosphorylation, trafficking, and processing of APP and apoEr2. Fyn expression has been shown to be significantly increased in Chronic Myelogenous Leukemia (CML). Knockdown of Fyn with shRNA slows leukemia cell growth, inhibits clonogenicity, and leads to increased sensitivity to imatinib.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Spezifikation
SLK, SYN, MGC45350 | |
Human | |
Recombinant | |
GST Tag |