missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
FoxH1 Polyclonal antibody specifically detects FoxH1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Spezifikation
Spezifikation
| Antigen | FoxH1 |
| Anwendungen | Western Blot, ELISA, Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugat | Alexa Fluor 647 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | Fast-1, FAST1forkhead box protein H1, Fast-2, FAST2, Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, forkhead activin signal transducer-1, forkhead box H1, hFAST-1, TGF-beta/activin signal transducer |
| Wirtsspezies | Rabbit |
| Immunogen | Antibody was raised against a 19 amino acid peptide near the amino terminus of human FOXH1. The immunogen is located within the first 50 amino acids of FOXH1. Amino Acid Squence: CQPPKRRKKRYLRHDKPPYT |
| Reinigungsverfahren | Peptide affinity purified |
| Menge | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?