Learn More
Finasteride, Thermo Scientific™
Inhibitor of steroid 5-a-reductase
Marke: Thermo Scientific Alfa Aesar J63454.03
| Menge | 1g |
|---|
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsFinasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.
Solubility
Soluble in water, DMSO, ethanol, methanol, and chloroform.
Notes
Protect from heat. Store away from oxidizing agents.
Chemische Identifikatoren
| 98319-26-7 | |
| 372.553 | |
| DBEPLOCGEIEOCV-WSBQPABSSA-N | |
| 57363 | |
| (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide |
| C23H36N2O2 | |
| MFCD00869737 | |
| finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin | |
| CHEBI:5062 | |
| CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C |
Spezifikation
| Finasteride | |
| C23H36N2O2 | |
| 1g | |
| finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin | |
| DBEPLOCGEIEOCV-WSBQPABSSA-N | |
| (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide | |
| 57363 | |
| 372.54 |
| 98319-26-7 | |
| MFCD00869737 | |
| 14,4082 | |
| Soluble in water,DMSO,ethanol,methanol,and chloroform. | |
| CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C | |
| 372.553 | |
| CHEBI:5062 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.