Learn More
Invitrogen™ FGFR4 Recombinant Human Protein
FGFR4 is a member of the FGFR family of receptor tyrosine kinases.
Brand: Invitrogen™ PR4380A
Additional Details : Gewicht : 0.00900kg
Description
The FGFR family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.
Specifications
NP_002002 | |
50mM Tris, pH 7.5, 100mM NaCl, 1mM DTT, 0.01% NP-40, 0.01mM EDTA, 20% glycerol | |
1mg | |
RUO - research use only | |
FGFR4, JTK2 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
42.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
FGF Receptor Kinase Family | |
Recombinant | |
Baculovirus |