missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR1OP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11343-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a largely hydrophilic protein postulated to be a leucine-rich protein family member. A t(6;8)(q27;p11) chromosomal translocation, fusing this gene and the fibroblast growth factor receptor 1 (FGFR1) gene, has been found in cases of myeloproliferative disorder. The resulting chimeric protein contains the N-terminal leucine-rich region of this encoded protein fused to the catalytic domain of FGFR1. This gene is thought to play an important role in normal proliferation and differentiation of the erythroid lineage. Alternatively spliced transcript variants that encode different proteins have been identified.Spezifikation
FGFR1OP | |
Polyclonal | |
Unconjugated | |
FGFR1OP | |
FGFR1 oncogene partner, FGFR1OP, FOP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11116, 683722, 75296 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95684, Q4V7C1, Q66JX5 | |
Fgfr1op | |
FGFR1OP Fusion Protein Ag1891 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |