Learn More
Invitrogen™ FGF1 (FGF alpha) Human ELISA Kit
Sandwich ELISA Kit
800.00 CHF - 5150.00 CHF
Spezifikation
Zugriffsnummer | Gehäuse P05230 |
---|---|
Assay-Empfindlichkeit: | 12 pg/mL |
Assay | Festphasen-Sandwich-ELISA |
Konjugat | Biotin |
Produkttyp | ELISA-Kit |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15404993
|
Invitrogen™
EHFGF1 |
96 Test-Kit |
800.00 CHF
1 Set |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
17854200
|
Invitrogen™
EHFGF1X10 |
10 x 96 Tests |
5150.00 CHF
10 Stück |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
17155439
|
Invitrogen™
EHFGF1X5 |
5 x 96 Tests |
2895.00 CHF
1 Stück |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The Human Fibroblast Growth Factor 1 (Hu FGF1/FGFα) ELISA quantitates Hu FGF1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FGF1. Principle of the method The Human FGF1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis. Multiple alternatively spliced variants encoding different isoforms have been described.Spezifikation
Gehäuse P05230 | |
Festphasen-Sandwich-ELISA | |
ELISA-Kit | |
Plasma, Serum, Überstand | |
Human | |
Kolorimetrisches Mikrotiterplatten-Lesegerät | |
AFGF,ECGF,ECGF-beta,ECGFA,ECGFB,FGF-1,FGF-alpha,FGFA,GLIO703,HBGF-1,HBGF1 | |
< 10 % | |
RUO | |
2 bis 8 °C | |
Human | |
4 Std 45 Min |
12 pg/mL | |
Biotin | |
Wachstumsfaktoren und Rezeptoren | |
Dieses ELISA-Kit zeigt keine Kreuzreaktivität mit einem der folgenden getesteten Zytokine: Humanes BLC, BDNF, bFGF, BMP-4, BMP-5, BMP-7, b-NGF, Eotaxin-1, Eotaxin-2, FGF-4, FGF-7, G-CSF, GM-CSF, I-309, ICAM-1, IFN-gamma, IL-1 alpha, IL-1 beta, IL-1 ra, IL-2, IL-4, IL-5, IL-6, IL-6 sR, IL-7, IL-8, IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-16, IL-17, MCP-1, M-CSF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PDGF-BB, RANTES, sTNFRI, TIMP-1, TIMP-2, TNF alpha, TNF beta, VEGF, VEGFR2, VEGFR3. | |
ELISA, Protein-Assays, Proteinbiologie, Krebsbiologie, Immunologie, Neurowissenschaften, Neurobiologie, Proteinnachweis, Proteinanalyse | |
2246 | |
< 12 % | |
HRP | |
Angiogenese | |
AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1 | |
1 Std 20 Min |