missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FER Recombinant Human Protein

FER is a member of the FPS/FES family of non-receptor tyrosine kinases.

Marke:  Invitrogen™ PR7343A

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15458563

  • 2605.00 CHF / 100 Mikrogramm
Verfügbar ab: 23-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

FER regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors.

Spezifikation

Spezifikation

NP_005237
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT and 50% glycerol
100μg
RUO - research use only
FER, TYK3
Human
His Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
60.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
FER Kinase Family
Recombinant
Baculovirus
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote