missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FER Recombinant Human Protein
FER is a member of the FPS/FES family of non-receptor tyrosine kinases.
2605.00 CHF - 11220.00 CHF
Spezifikation
Zugriffsnummer | NP_005237 |
---|---|
Zur Verwendung mit (Anwendung) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Zusammensetzung | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT and 50% glycerol |
Molekulargewicht | 60.3kDa |
Lagerungsbedingungen | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15458563
|
Invitrogen™
PR7343A |
100μg |
2605.00 CHF
100 Mikrogramm |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
10452523
|
Invitrogen™
PV4076 |
1mg |
11220.00 CHF
1 Milligramm |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
FER regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors.
Spezifikation
NP_005237 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT and 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
FER Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
60.3kDa | |
RUO - research use only | |
FER, TYK3 | |
Human | |
His Tag | |
Frozen |