missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12802-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.Spezifikation
FABP4 | |
Polyclonal | |
Unconjugated | |
FABP4 | |
A FABP, AFABP, ALBP, aP2, FABP4, Fatty acid binding protein 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11770, 2167, 79451 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04117, P15090, P70623 | |
Fabp4 | |
FABP4 Fusion Protein Ag3912 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |