missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | FABP4 |
---|---|
Konzentration | 0.2 mg/mL |
Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850714
|
Proteintech
12802-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840714
|
Proteintech
12802-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.Spezifikation
FABP4 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P04117, P15090, P70623 | |
11770, 2167, 79451 | |
FABP4 Fusion Protein Ag3912 | |
Primary | |
-20°C | |
FABP4 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A FABP, AFABP, ALBP, aP2, FABP4, Fatty acid binding protein 4 | |
Fabp4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |