Learn More
EXO1 Mouse, anti-Human, Clone: 2D11D1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 68536-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The EXO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EXO1. This antibody recognizes human antigen. The EXO1 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| EXO1 | |
| 2D11D1 | |
| EXO1, EXOI, exonuclease 1, Exonuclease I, HEX1, hExo1, hExoI | |
| 20 μL | |
| 9156 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |