Learn More
Novus Biologicals™ Europium Streptavidin MINI
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
80 μl
Packungsgröße:
80 Mikroliter
Beschreibung
- Highly stable, easy to use conjugates
- Resistant to aggregation
- No sonication, shaking or vortexing required
- No need for blocking solution
- Possibility to work with low conjugate concentrations
- Conjugates can be run on an immunochromatographic assay as well as a microwell-based assay
Kit Contents:
- 1 Mini vial of 200 nmEuropium - Streptavidin conjugate.
Spezifikation
Spezifikation
| Menge | 80 μl |
| Inhalt und Lagerung | Storage of components varies. See protocol for specific instructions. |