missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRB Mouse anti-Human, Clone: 3E10C1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66818-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ESRRB (steroid hormone receptor ERR2) plays a role in maintaice of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survial of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).Spezifikation
ESRRB | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95718 | |
ESRRB | |
ESRRB Fusion Protein Ag18626 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
3E10C1 | |
Unconjugated | |
ESRRB | |
DFNB35, ERR2, ERRb, ERRbeta, ERRbeta 2, ESRL2, ESRRB, ESRRB protein, estrogen related receptor beta, NR3B2 | |
Mouse | |
Protein G | |
RUO | |
2103 | |
-20°C | |
Liquid |