missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRB Mouse anti-Human, Clone: 3E10C1, Proteintech
Mouse Monoclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | ESRRB |
---|---|
Klon | 3E10C1 |
Konzentration | 1 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16849045
|
Proteintech
66818-1-IG-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16839045
|
Proteintech
66818-1-IG-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
ESRRB (steroid hormone receptor ERR2) plays a role in maintaice of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survial of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).Spezifikation
ESRRB | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DFNB35, ERR2, ERRb, ERRbeta, ERRbeta 2, ESRL2, ESRRB, ESRRB protein, estrogen related receptor beta, NR3B2 | |
ESRRB | |
IgG1 | |
Protein G | |
Antibody |
3E10C1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O95718 | |
2103 | |
ESRRB Fusion Protein Ag18626 | |
Primary | |
-20°C | |
ESRRB |