Learn More
Epcam / CD326 Mouse, Clone: rMOC-31, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 4072-MSM19-P0
Additional Details : Gewicht : 0.01000kg
Spezifikation
Epcam / CD326 | |
Cell surface | |
rMOC-31 | |
Unconjugated | |
NP_002345 | |
Mouse | |
40-43kDa | |
Protein A/G Purified | |
RUO | |
4072 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
EPCAM | |
Neuraminidase treated GLS-1 human small cell lung carcinoma cells | |
HT29 cells or breast tumor | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |