Learn More
Merck Millipak™ Einweg-Filtereinsätze
Entfernt Partikel und Mikroorganismen aus Flüssigkeiten und Gasen mit einem pumpenangetriebenen Filter. Merck Millipore™ Millipak™ Einweg-Filtereinsätze bieten die sterile Filtration von Parenteralia, Diagnostika, Puffern, Medien und biologischen Proben.
Marke: Merck Chemicals MPVL02GH2
Additional Details : Gewicht : 0.20000kg
Beschreibung
Entfernt Partikel und Mikroorganismen aus Flüssigkeiten und Gasen mit einem pumpenangetriebenen Filter.Millipak™ Einweg-Filtereinsätze bieten die sterile Filtration von Parenteralia, Diagnostika, Puffern, Medien und biologischen Proben.
Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.
- Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
- Point-of-use filter for high-purity water
- Ideally suited for high value-added applications
- Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
- Presterilized units are gamma irradiated
- Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
- 100% integrity tested to guarantee integrity
Spezifikation
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | |
Polycarbonate | |
0.1 μm | |
100 cm2 | |
Removal of particles and microorganisms from liquids and gases | |
2/Pk. | |
10 L | |
8.4 cm | |
7.6 cm |
Single-use | |
PVDF | |
Sterile | |
0.7 L/min. at 1.75 bar | |
75 psi at 25°C | |
Filter Unit | |
3.3 in. | |
3 in. |