missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EID1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11734-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
E1A-like inhibitor of differentiation-1 (EID-1), an acetyltransferase enzyme, binds both the retinoblastoma protein (Rb), a regulator of cell cycle and tissue specific transcription, and the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. EID-1 inhibits cellular differentiation by blocking the histone acetyltransferase activity of p300. EID-1 also acetylates both histones and non-histone proteins such as NCOA3 co-activator. By acetylating histones, EID-1 gives a specific tag for transcriptional activation. In addition to binding Rb and p300, EID-1 also binds to phosphorylated CREB protein, mediating cAMP gene regulation. EID-1 augments the activity of phosphorylated CREB and activates transcription of cAMP responsive genes as a co-activator.Spezifikation
EID1 | |
Polyclonal | |
Unconjugated | |
EID1 | |
21 kDa pRb associated protein, C15orf3, CRI1, EID 1, EID1, IRO45620, PNAS 22, PTD014, RBP21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23741, 58521 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9DCR4, Q9Y6B2 | |
Eid1 | |
EID1 Fusion Protein Ag2336 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |