Learn More
Duran™ Dispenser
Marke: Duran™ 291150908
Additional Details : Gewicht : 0.17400kg
Spezifikation
<±0.6 to 0.5% | |
Autoclavable at 121°C | |
1 to 10 mL | |
<± 0.1% |
Bottletop Dispenser | |
DIN ISO 8655 | |
GL 28, GL 45, S 40 Adapters |