missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIRAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12634-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
DIRAS1 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.Spezifikation
DIRAS1 | |
Polyclonal | |
Unconjugated | |
DIRAS1 | |
Di Ras1, DIRAS1, GBTS1, GTP binding protein Di Ras1, RIG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
148252, 208666, 366826 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95057, Q91Z61 | |
Diras1 | |
DIRAS1 Fusion Protein Ag3329 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |