Learn More
DICER1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48 CHF - 423.57 CHF
Spezifikation
| Antigen | DICER1 |
|---|---|
| Konzentration | 1000 μg/mL |
| Anwendungen | Immunofluorescence |
| Konjugat | CoraLite Plus 488 |
| Form | Liquid |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18077105
|
Proteintech
CL488-20567-20UL |
20 μL |
136.48 CHF
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18015025
|
Proteintech
CL488-20567-100UL |
100 μL |
423.57 CHF
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The DICER1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human DICER1. This antibody recognizes human antigen. The DICER1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Spezifikation
| DICER1 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| DCR1, Dicer, dicer 1, ribonuclease type III, DICER1, Endoribonuclease Dicer, Helicase MOI, Helicase with RNase motif, HERNA, KIAA0928 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 23405 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |