Learn More
DICER1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL488-20567-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The DICER1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human DICER1. This antibody recognizes human antigen. The DICER1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Spezifikation
| DICER1 | |
| 1000 μg/mL | |
| DCR1, Dicer, dicer 1, ribonuclease type III, DICER1, Endoribonuclease Dicer, Helicase MOI, Helicase with RNase motif, HERNA, KIAA0928 | |
| 100 μL | |
| 23405 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |