Learn More
DGK-beta Antibody (OTI1F9), DyLight 680, Novus Biologicals™
Artikelnummer. 18108232 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18108232 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18108232 Lieferant Novus Biologicals Lieferanten-Nr. NBP270552FR
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Monoclonal Antibody
DGK-beta Monoclonal specifically detects DGK-beta in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen DGK-beta
Anwendungen Western Blot
Klassifikation Monoclonal
Klon OTI1F9
Konjugat DyLight 680
Verdünnung Western Blot
Zusammensetzung PBS with 0.05% Sodium Azide
Gen-Alias 90 kDa diacylglycerol kinase, DAG kinase beta, DAGK2, DGK-beta, diacylglycerol kinase beta, diacylglycerol kinase, beta (90kD), diacylglycerol kinase, beta 90kDa, Diglyceride kinase beta, EC 2.7.1, EC 2.7.1.107, KIAA0718DGKDGK-BETA
Gensymbole DGKB
Wirtsspezies Mouse
Immunogen Full length recombinant protein of human DGK-beta (NP_663733) produced in HEK293T cells.
Reinigungsverfahren Affinity Purified
Menge 0.1 mL
Forschungsgebiet Lipid and Metabolism
Primär oder sekundär Primary
Gen-ID (Entrez) 1607.0
Zielspezies Human, Mouse, Rat, Canine, Monkey, Primate
Inhalt und Lagerung Store at 4°C in the dark.
Isotype IgG1