missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEAF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18323-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
HNF1A is a transcription factor required for the expression of several liver-specific genes. This protein functions as a homodimer and binds to the inverted palindrome 5′-GTTAATNATTAAC-3′.Spezifikation
DEAF1 | |
Polyclonal | |
Unconjugated | |
DEAF1 | |
DEAF1, NUDR, SPN, Suppressin, ZMYND5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10522, 54006, 83632 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75398, O88450, Q9Z1T5 | |
Deaf1 | |
DEAF1 Fusion Protein Ag13173 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |