Learn More
DC-SIGNR/CD299/CLEC4M Antibody (16E7), DyLight 680 - Chimeric, Novus Biologicals™
Artikelnummer. 18759692 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18759692 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18759692 Lieferant Novus Biologicals Lieferanten-Nr. NBP252649FR
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Monoclonal Antibody
DC-SIGNR/CD299/CLEC4M Monoclonal antibody specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen DC-SIGNR/CD299/CLEC4M
Anwendungen Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, CyTOF
Klassifikation Monoclonal
Klon 16E7
Konjugat DyLight 680
Zusammensetzung 50 mM Sodium Borate
Gen-Alias CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR
Wirtsspezies Rabbit
Immunogen CD299
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Signal Transduction
Primär oder sekundär Primary
Gen-ID (Entrez) 10332
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG κ