Learn More
DC-SIGN/CD209 Antibody (MMD3), mFluor Violet 500 SE, Novus Biologicals™
Artikelnummer. 18600170 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18600170 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18600170 Lieferant Novus Biologicals Lieferanten-Nr. FAB83451MFV500
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Monoclonal Antibody
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen DC-SIGN/CD209
Anwendungen Flow Cytometry, CyTOF
Klassifikation Monoclonal
Klon MMD3
Konjugat mFluor Violet 500 SE
Verdünnung Flow Cytometry, CyTOF-ready
Zusammensetzung 50mM Sodium Borate
Gen-Alias CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Wirtsspezies Mouse
Immunogen Purified mouse DC-SIGN/CD209 extracellular domain, Accession # FJ168685
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primär oder sekundär Primary
Gen-ID (Entrez) 30835
Zielspezies Mouse
Inhalt und Lagerung Store at 4C in the dark.
Isotype IgG2c