missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DAZ1 Polyclonal antibody specifically detects DAZ1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | DAZ1 |
| Anwendungen | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Zusammensetzung | 0.2 μm filtered solution in PBS |
| Gen-Alias | deleted in azoospermia, deleted in azoospermia 1, FLJ17175, SPGYDAZdeleted in azoospermia protein 1 |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DAZ1. |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?