missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DAPK2, His Tag Recombinant Protein
DAPK2 Recombinant Human Protein
842.00 CHF
Spezifikation
Zugriffsnummer | NP_055141 |
---|---|
Zur Verwendung mit (Anwendung) | Kinase Assay |
Molekulargewicht | 47.9 kDa |
Gen-Alias | DAPK2, DRP-1 |
Kinase-Familie | DAPK Kinase Family |
Beschreibung
DAPK2 is a calcium⁄calmodulin-dependent kinase that acts as a positive regulator of apoptosis, and is expressed in many tissue types.Spezifikation
NP_055141 | |
47.9 kDa | |
DAPK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 300 mM NaCl, 0.5 mM EDTA, 0.05% Brij-35, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |
Kinase Assay | |
DAPK2, DRP-1 | |
Human | |
His Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus |