missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DAPK2, His Tag Recombinant Protein
DAPK2 Recombinant Human Protein
Marke: Invitrogen PV3614
Additional Details : Gewicht : 0.05250kg
Beschreibung
DAPK2 is a calcium⁄calmodulin-dependent kinase that acts as a positive regulator of apoptosis, and is expressed in many tissue types.Spezifikation
NP_055141 | |
23604 | |
10 μg | |
DAPK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 300 mM NaCl, 0.5 mM EDTA, 0.05% Brij-35, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CAMK Group | |
DAPK2 | |
Baculovirus | |
Recombinant, Full Length | |
Signal Transduction, Apoptosis |
Kinase Assay | |
47.9 kDa | |
DAPK2, DRP-1 | |
Human | |
His Tag | |
Biochemical binding/kinetic | |
Dry Ice | |
death-associated protein kinase 2 | |
Kinases & Inhibitors | |
Serine/Threonine Kinases |