missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAPK1 Recombinant Human Protein
DAPK1 is a large, calmodulin-dependent kinase.
Marke: Invitrogen™ PV4273
Additional Details : Gewicht : 0.00900kg
Beschreibung
DAPK1 is involved in gamma-interferon induced programmed cell death and is a potential tumour suppressor.
Spezifikation
NP_004929 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
DAPK1 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
71.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
DAPK Kinase Family | |
Recombinant | |
Baculovirus |