missing translation for 'onlineSavingsMsg'
Learn More

D-Glucurono-6,3-lactone acetonide, 98+%

Product Code. 11304755
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11304755

Brand: Thermo Scientific Alfa Aesar L17765.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20513-98-8 | C9H12O6 | 216.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20513-98-8
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
MDL Number MFCD00061641
InChI Key BDBGJSXZKMTMGP-BOFVIFSBSA-N
Synonym d-glucurono-6,3-lactone acetonide
PubChem CID 134129430
IUPAC Name (3aS,3bS,6S,6aS,7aR)-6-hydroxy-2,2-dimethyl-3b,6,6a,7a-tetrahydro-3aH-furo[2,3]furo[2,4-b][1,3]dioxol-5-one
SMILES CC1(OC2C3C(C(C(=O)O3)O)OC2O1)C

Specifications

Melting Point 119°C to 121°C
Quantity 50 g
Optical Rotation +52° (c=1 in Chloroform)
Formula Weight 216.19
Percent Purity ≥98%
Chemical Name or Material D-Glucurono-6,3-lactone acetonide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.