missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 13 Mouse anti-Human, Rat, Clone: 1E10G2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66684-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Cytokeratin 13 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This type I cytokeratin is paired with keratin 4 and expressed in the suprabasal layers of non-cornified stratified epithelia. Mutations in this gene and keratin 4 have been associated with the autosomal domit disorder White Sponge Nevus. The type I cytokeratins are clustered in a region of chromosome 17q21.2. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been described.Spezifikation
Cytokeratin 13 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13646, Q6IFV4 | |
Krt13 | |
Cytokeratin 13 Fusion Protein Ag0217 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1E10G2 | |
Unconjugated | |
KRT13 | |
CK 13, CK13, Cytokeratin 13, K13, keratin 13, KRT13 | |
Mouse | |
Protein G | |
RUO | |
287699, 3860 | |
-20°C | |
Liquid |