Learn More
ACROBiosystems Cynomolgus CRTAM Protein, Fc Tag
365.69 CHF - 2522.00 CHF
Spezifikation
Zugriffsnummer | XP_005580021.1 |
---|---|
Molekulargewicht | 56.6 kDa |
Quelle | Cynomolgus CRTAM, Fc Tag (CRM-C5255) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Gly 287 (Accession # XP_005580021.1). |
Immunogen | Ser 18 - Gly 287 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15816477
|
ACROBiosystems
CRM-C5255-100UG |
100 μg |
365.69 CHF
100 Mikrogramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826477
|
ACROBiosystems
CRM-C5255-1MG |
1 mg |
2522.00 CHF
1 Milligramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Class I-restricted T cell-associated molecule (CRTAM), a member of nectin family and the immunoglobulin superfamily, is also known as cytotoxic and regulatory T-cell molecule, which is expressed by activated CD8+ and NK T cells. CRTAM is found in spleen, thymus, small intestine, peripheral blood, and surprisingly, in brain where it is highly expressed by Purkinje cells of the cerebellum. The high affinity of CRTAM/IGSF4 adhesion allows CRTAM to disrupt IGSF4 homotypic interactions (3 - 5). IGSF4 and T cell receptor co-engagement of CRTAM-expressing CD8+ cells induces increased IFN-gamma or IL-22 production (3, 4). Furthermore, a role in cancer surveillance through NK cell-mediated rejection of IGSF4-expressing tumors has been proposed.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.6 kDa. The protein migrates as 66-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
XP_005580021.1 | |
Cynomolgus CRTAM, Fc Tag (CRM-C5255) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Gly 287 (Accession # XP_005580021.1). | |
-20°C | |
CRTAM | |
Cynomolgus Monkey | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
56.6 kDa | |
Ser 18 - Gly 287 | |
CRTAM,CD355 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |