Learn More
ACROBiosystems Cynomolgus CRTAM Protein, Fc Tag
Marke: ACROBiosystems CRM-C5255-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Class I-restricted T cell-associated molecule (CRTAM), a member of nectin family and the immunoglobulin superfamily, is also known as cytotoxic and regulatory T-cell molecule, which is expressed by activated CD8+ and NK T cells. CRTAM is found in spleen, thymus, small intestine, peripheral blood, and surprisingly, in brain where it is highly expressed by Purkinje cells of the cerebellum. The high affinity of CRTAM/IGSF4 adhesion allows CRTAM to disrupt IGSF4 homotypic interactions (3 - 5). IGSF4 and T cell receptor co-engagement of CRTAM-expressing CD8+ cells induces increased IFN-gamma or IL-22 production (3, 4). Furthermore, a role in cancer surveillance through NK cell-mediated rejection of IGSF4-expressing tumors has been proposed.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.6 kDa. The protein migrates as 66-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
XP_005580021.1 | |
1 mg | |
Ser 18 - Gly 287 | |
CRTAM,CD355 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
56.6 kDa | |
Cynomolgus CRTAM, Fc Tag (CRM-C5255) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Gly 287 (Accession # XP_005580021.1). | |
-20°C | |
CRTAM | |
Cynomolgus Monkey | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |