Learn More
CXCR5 Rat anti-Mouse, PE, Clone: 2G8, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561988
Additional Details : Gewicht : 0.00800kg
Beschreibung
The 2G8 monoclonal antibody specifically binds to the mouse CXC chemokine receptor, CXCR5. CXCR5 is also known as CD185, BLR1, NLR and MDR15. CXCR5 is a seven-transmembrane, G-protein-coupled receptor that is specific for the CXC chemokine, CXCL13/BLC/BCA-1. The expression of CXCR5 has been detected in spleen, lymph nodes, tonsils, brain, bone marrow, T cells, B cells, cerebrum, cerebellum, hippcampus and pituitary. In mouse spleen, CXCR5 was strictly expressed by mature B cells and a small subset of T lymphocytes. CXCR5 plays a role in directing the migration of B and T cells to B cell follicles with the spleen and certain other lymphoid tissues. The immunogen used to generate 2G8 hybridoma was a recombinant protein containing N-terminal amino acids of mouse CXCR5 (GST-NmBLR1).
Spezifikation
CXCR5 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse CXCR5 | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
2G8 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
For Research Use Only.