Beschreibung
CXCR3 Polyclonal antibody specifically detects CXCR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Spezifikation
Spezifikation
| Antigen | CXCR3 |
| Anwendungen | Western Blot, ELISA, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konjugat | mFluor Violet 450 SE |
| Verdünnung | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR |
| Wirtsspezies | Rabbit |
| Immunogen | Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human CXCR3 . The immunogen is located within the last 50 amino acids of CXCR3. Amino Acid Squence: RGLQRQPSSSRRDSSWSET |
| Reinigungsverfahren | Peptide affinity purified |
| Mehr anzeigen |