Beschreibung
CPSF6 Polyclonal specifically detects CPSF6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Immunohistochemistry (Paraffin)/Immunoprecipitation/Western Blot.
Spezifikation
Spezifikation
| Anwendungen | Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Konjugat | PE |
| Wirtsspezies | Rabbit |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Inhalt und Lagerung | Store at 4C in the dark. |
| Produkttyp | Antibody |
| Form | Purified |
| Isotype | IgG |
| Produktlinie | Novus |