Learn More
CoraLite™ Plus 488-conjugated Syntaxin 6 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
130.95 CHF - 420.90 CHF
Spezifikation
| Antigen | Syntaxin 6 |
|---|---|
| Klon | 2B6G4 |
| Konzentration | 1000 μg/mL |
| Anwendungen | Immunofluorescence |
| Konjugat | CoraLite Plus 488 |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30235420
|
Proteintech
CL488-60059-100UL |
100 μL |
420.90 CHF
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30237825
|
Proteintech
CL488-60059-20UL |
20 μL |
130.95 CHF
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The Syntaxin 6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Syntaxin 6. This antibody recognizes human, mouse, rat antigen. The Syntaxin 6 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Spezifikation
| Syntaxin 6 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 10228 | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
| 2B6G4 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |