Learn More
CoraLite™ Plus 488-conjugated Syntaxin 6 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-60059-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Syntaxin 6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Syntaxin 6. This antibody recognizes human, mouse, rat antigen. The Syntaxin 6 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Spezifikation
| Syntaxin 6 | |
| 2B6G4 | |
| CoraLite Plus 488 | |
| Recombinant Protein | |
| 100 μL | |
| 10228 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rat | |
| 29 kDa | |
| Liquid |