Learn More
BOLA Conductors for Magnetic Stirrer Heads
Click to view available options
Typ:
Haltestange/Befestigungsstange
Packungsgröße:
1 Stück
Beschreibung
- Borosilicate glass with ground joint and glued upper bearing.
- Alternatively without ground joint for fusing on the reactor lid
- Upper bearing to be glued is included with Ground Joint NS 29/32
Spezifikation
Spezifikation
| Typ | Haltestange/Befestigungsstange |
| Material | Glass |