missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMP Mouse anti-Human, Porcine, Clone: 1D7A6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66793-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
COMP is a noncollagenous extracellular matrix (ECM) protein. It consists of five identical glycoprotein subunits, each with EGF like and calcium binding (thrombospondin like) domains. Oligomerization results from formation of a five stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED).Spezifikation
COMP | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49747 | |
COMP | |
COMP Fusion Protein Ag4575 | |
20 μL | |
Primary | |
Porcine, Human | |
Antibody | |
IgG1 |
Western Blot | |
1D7A6 | |
Unconjugated | |
COMP | |
COMP, EDM1, EPD1, MED, PSACH, THBS5, Thrombospondin 5, TSP5 | |
Mouse | |
Protein G | |
RUO | |
1311, 396554 | |
-20°C | |
Liquid |