Learn More
Novus Biologicals™ Colon Matched Tumor and Normal Tissue Lysate
Artikelnummer. 18104188 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
2 Fläschchen
Packungsgröße:
2 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18104188 2 Fläschchen 2 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18104188 Lieferant Novus Biologicals™ Lieferanten-Nr. NBP247079
  • ONLINE EXCLUSIVE PRICE
    580.00 CHF
    548.10 CHF / 2 Stück
    Sparen 31.90 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 06-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Two matched 1mg/mL vials (100μg/vial) of human colon tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Colon Matched Tumor & Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Spezifikation
Tissue Colon
Menge 2 Fläschchen
Wirtsspezies Human
Inhalt und Lagerung Store at −80°C. Avoid freeze-thaw cycles.