missing translation for 'onlineSavingsMsg'
Learn More

Collagen I Antibody, R&D Systems™

Artikelnummer. 18070976 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0.025 mg
Packungsgröße:
25 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18070976 0.025 mg 25 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18070976 Lieferant Novus Biologicals Lieferanten-Nr. NB6004080.025MG
 Eingeschränkter Artikel
Verfügbar ab: 23-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody has been used in 195 publications

Collagen I alpha 1 Polyclonal specifically detects Collagen I alpha 1 in Bacteria, Bovine, Chicken, Human, Mouse, Pig, Rabbit, Rat, Zebrafish samples. It is validated for ELISA, Fluorophore-linked immunosorbent assay, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount, Immunoprecipitation, KnockDown, Microarray, SDS-Page, Western Blot.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Collagen I alpha 1
Anwendungen ELISA, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Immunoprecipitation, KnockDown, Microarray, SDS-Page, Western Blot
Klassifikation Polyclonal
Konzentration 1.0 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1:1000, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry 1:1600
Zusammensetzung 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gen-Zugriffsnummer P02452, P02453
Gen-Alias Alpha-1 type I collagen, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, Collagen1, Collagen-1, OI4, pro-alpha-1 collagen type 1
Gensymbole COL1A1
Wirtsspezies Rabbit
Immunogen Collagen Type I from human and bovine placenta by limited pepsin digestion and selective salt precipitation.
Molekulargewicht des Antigens 139 kDa
Reinigungsverfahren Affinity Purified
Menge 0.025 mg
Regulatorischer Status RUO
Forschungsgebiet Cancer, Cell Cycle and Replication, Cytoskeleton Markers, Extracellular Matrix
Primär oder sekundär Primary
Gen-ID (Entrez) 1277
Testspezifität Typically less than 1% cross-reactivity against other types of collagens was detected by ELISA against purified standards. Some class-specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Type I collagens and has negligible cross-reactivity with Type II, III, IV, V or VI collagens. Non-specific cross-reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible.
Zielspezies Bacteria, Bovine, Chicken, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Produkttyp Antibody
Isotype IgG
Produktlinie Novus
Mehr anzeigen Weniger anzeigen

For Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.