missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL1A2 Mouse anti-Human, Porcine, Clone: 3D5E8, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66761-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Type I collagen is a member of group I collagen (fibrillar forming collagen).Spezifikation
COL1A2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08123 | |
COL1A2 | |
COL1A2 Fusion Protein Ag6281 | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
3D5E8 | |
Unconjugated | |
COL1A2 | |
Alpha 2 type I collagen, COL1A2, Collagen alpha 2(I) chain, collagen, type I, alpha 2, OI4 | |
Mouse | |
Protein G | |
RUO | |
100626716, 1278 | |
-20°C | |
Liquid |