missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11848-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus.Spezifikation
Cofilin 2 | |
Polyclonal | |
Unconjugated | |
CFL2 | |
CFL2, Cofilin 2, cofilin 2 (muscle), Cofilin, muscle isoform, NEM7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1073, 12632, 366624 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P45591, Q9Y281 | |
Cfl2 | |
Cofilin 2 Fusion Protein Ag2408 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |